siRNA Details
| Gene Symbol: Gxylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100887.1 | NP_001094357.1 | 3 | 437 | ||
| XM_006242192.2 | XP_006242254.1 | 2 | 345 | |||
| GenBank mRNA | AY623036.1 | - | 1 | 40 | ||
| BC167068.1 | - | 2 | 191 | 8363586 | ||

Gxylt1
Genomic Map