siRNA Details
| Gene Symbol: Lmbr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013950.1 | NP_001013972.1 | 8 | 798 | ||
| XM_008765698.1 | XP_008763920.1 | 8 | 845 | |||
| XM_008765699.1 | XP_008763921.1 | 8 | 841 | |||
| XM_008765700.1 | XP_008763922.1 | 6 | 730 | |||
| GenBank mRNA | BC089929.1 | - | 8 | 798 | 7308748 | |

Lmbr1l
Genomic Map