siRNA Details
| Gene Symbol: Eif4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008324.1 | NP_001008325.1 | 9 | 1010 | ||
| GenBank mRNA | BC085933.1 | - | 9 | 1010 | 7112408 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif4b| Gene Symbol: Eif4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008324.1 | NP_001008325.1 | 9 | 1010 | ||
| GenBank mRNA | BC085933.1 | - | 9 | 1010 | 7112408 | |