siRNA Details
| Gene Symbol: Sp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037632.1 | NP_001032721.1 | 2 | 984 | ||
| NM_181374.2 | NP_852039.2 | 2 | 864 | |||
| XM_006242390.2 | XP_006242452.1 | 2 | 1220 | |||
| XM_008765706.1 | XP_008763928.1 | 3 | 790 | |||
| GenBank mRNA | AY177399.1 | - | 2 | 766 | ||

Sp7
Genomic Map