siRNA Details
| Gene Symbol: Alg13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013951.1 | NP_001013973.1 | 1 | 45 | ||
| GenBank mRNA | BC088233.1 | - | 1 | 45 | 7324702 | |
| FQ216283.1 | - | 1 | 57 | |||
| FQ229089.1 | - | 1 | 71 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alg13| Gene Symbol: Alg13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013951.1 | NP_001013973.1 | 1 | 45 | ||
| GenBank mRNA | BC088233.1 | - | 1 | 45 | 7324702 | |
| FQ216283.1 | - | 1 | 57 | |||
| FQ229089.1 | - | 1 | 71 | |||