siRNA Details
| Gene Symbol: Kctd17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134529.1 | NP_001128001.1 | 5, 6 | 671 | ||
| XM_006241976.2 | XP_006242038.1 | 5, 6 | 817 | |||
| XM_006241977.2 | XP_006242039.1 | 5, 6 | 811 | |||
| XM_008765708.1 | XP_008763930.1 | 5, 6 | 814 | |||
| XM_008765709.1 | XP_008763931.1 | 5, 6 | 812 | |||
| XM_008765710.1 | XP_008763932.1 | 5, 6 | 815 | |||

Kctd17
Genomic Map