siRNA Details
| Gene Symbol: Mbd3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106803.1 | NP_001100273.1 | 3 | 172 | ||
| XM_006242571.2 | XP_006242633.1 | 2 | 257 | |||
| XM_006242574.2 | XP_006242636.1 | 3 | 533 | |||
| XM_006242576.2 | XP_006242638.1 | 2 | 88 | |||

Mbd3l1
Genomic Map