siRNA Details
| Gene Symbol: Tmem205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106804.1 | NP_001100274.1 | 3 | 969 | ||
| XM_006242621.2 | XP_006242683.1 | 3 | 398 | |||
| XM_006242622.2 | XP_006242684.1 | 3 | 240 | |||
| XM_006242623.2 | XP_006242685.1 | 4 | 314 | |||
| XM_006242624.2 | XP_006242686.1 | 4 | 579 | |||
| XM_006242625.2 | XP_006242687.1 | 3 | 286 | |||
| XM_008765928.1 | XP_008764150.1 | 4 | 303 | |||
| XM_008765929.1 | XP_008764151.1 | 3 | 281 | |||

Tmem205
Genomic Map