siRNA Details
| Gene Symbol: Tmem218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008325.1 | NP_001008326.1 | 3 | 201 | ||
| XM_006242802.2 | XP_006242864.1 | 3 | 316 | |||
| XM_008766050.1 | XP_008764272.1 | 3 | 352 | |||
| GenBank mRNA | BC085342.1 | - | 3 | 201 | 7303171 | |
| FQ214300.1 | - | 3 | 221 | |||

Tmem218
Genomic Map