siRNA Details
| Gene Symbol: Olr1293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000460.1 | NP_001000460.1 | 1 | 568 | ||
| Gene Symbol: LOC100910599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770723.1 | XP_008768945.1 | 1 | 568 | ||

Olr1293
Genomic Map