siRNA Details
Gene Symbol: Crtam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106813.1 | NP_001100283.1 | 6 | 586 | ||
XM_008766122.1 | XP_008764344.1 | 6 | 691 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Crtam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106813.1 | NP_001100283.1 | 6 | 586 | ||
XM_008766122.1 | XP_008764344.1 | 6 | 691 |