siRNA Details
| Gene Symbol: C2cd2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011996.1 | NP_001011996.1 | 9, 10 | 1560 | ||
| XM_006242917.2 | XP_006242979.1 | 9, 10 | 1611 | |||
| XM_006242918.2 | XP_006242980.1 | 9, 10 | 1611 | |||
| XM_006242919.2 | XP_006242981.1 | 9, 10 | 1611 | |||
| XM_006242920.2 | XP_006242982.1 | 9, 10 | 1611 | |||
| XM_008766156.1 | XP_008764378.1 | 9, 10 | 1611 | |||
| XM_008766157.1 | XP_008764379.1 | 9, 10 | 1163 | |||
| GenBank mRNA | BC085925.1 | - | 9, 10 | 1560 | 7109537 | |

C2cd2l
Genomic Map