siRNA Details
Gene Symbol: Cd3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077646.2 | NP_001071114.1 | 3 | 201 | ||
GenBank mRNA | S79711.1 | - | 3 | 201 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077646.2 | NP_001071114.1 | 3 | 201 | ||
GenBank mRNA | S79711.1 | - | 3 | 201 |