siRNA Details
Gene Symbol: Cd3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077646.2 | NP_001071114.1 | 4, 5 | 500 | ||
XM_006242932.2 | XP_006242994.1 | 4, 5 | 525 | |||
GenBank mRNA | S79711.1 | - | 4, 5 | 500 |
