siRNA Details
| Gene Symbol: Slc35f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106822.1 | NP_001100292.1 | 4 | 757 | ||
| XM_006243069.2 | XP_006243131.1 | 5 | 1001 | |||
| GenBank mRNA | BC091334.1 | - | 5 | 713 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35f2| Gene Symbol: Slc35f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106822.1 | NP_001100292.1 | 4 | 757 | ||
| XM_006243069.2 | XP_006243131.1 | 5 | 1001 | |||
| GenBank mRNA | BC091334.1 | - | 5 | 713 | ||