siRNA Details
| Gene Symbol: Cib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015010.1 | NP_001015010.1 | 1, 2 | 229 | ||
| GenBank mRNA | BC092630.1 | - | 1, 2 | 229 | 7314828 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cib2| Gene Symbol: Cib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015010.1 | NP_001015010.1 | 1, 2 | 229 | ||
| GenBank mRNA | BC092630.1 | - | 1, 2 | 229 | 7314828 | |