siRNA Details
Gene Symbol: Lman1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012465.1 | NP_001012483.1 | 1 | 126 | ||
GenBank mRNA | AB188302.1 | - | 1 | 126 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lman1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012465.1 | NP_001012483.1 | 1 | 126 | ||
GenBank mRNA | AB188302.1 | - | 1 | 126 |