siRNA Details
| Gene Symbol: Hexa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004443.1 | NP_001004443.1 | 2 | 343 | ||
| XM_006243162.1 | XP_006243224.1 | 3 | 213 | |||
| GenBank mRNA | BC082097.1 | - | 2 | 343 | 7134440 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hexa| Gene Symbol: Hexa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004443.1 | NP_001004443.1 | 2 | 343 | ||
| XM_006243162.1 | XP_006243224.1 | 3 | 213 | |||
| GenBank mRNA | BC082097.1 | - | 2 | 343 | 7134440 | |