siRNA Details
Gene Symbol: Coro2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_217183.8 | XP_217183.6 | 7 | 945 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Coro2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_217183.8 | XP_217183.6 | 7 | 945 |