siRNA Details
| Gene Symbol: Coro2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_217183.8 | XP_217183.6 | 11 | 1353 | ||
| GenBank mRNA | BC089991.1 | - | 1 | 21 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Coro2b
Coro2b| Gene Symbol: Coro2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_217183.8 | XP_217183.6 | 11 | 1353 | ||
| GenBank mRNA | BC089991.1 | - | 1 | 21 | ||
 Back To Top
 Back To Top