siRNA Details
| Gene Symbol: Dapk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013109.1 | NP_001013127.1 | 4 | 416 | ||
| XM_006243306.2 | XP_006243368.1 | 8 | 933 | |||
| XM_006243307.1 | XP_006243369.1 | 5 | 410 | |||
| XM_006243308.2 | XP_006243370.1 | 5 | 1612 | |||
| XM_006243309.2 | XP_006243371.1 | 5 | 1307 | |||
| XM_008766241.1 | XP_008764463.1 | 8 | 943 | |||
| GenBank mRNA | BC091410.1 | - | 4 | 416 | 7325495 | |

Dapk2
Genomic Map