siRNA Details
| Gene Symbol: Leo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005548.1 | NP_001005548.1 | 2, 3 | 856 | ||
| GenBank mRNA | BC082098.1 | - | 2, 3 | 856 | 7134452 | |
| FQ229860.1 | - | 2, 3 | 857 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Leo1| Gene Symbol: Leo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005548.1 | NP_001005548.1 | 2, 3 | 856 | ||
| GenBank mRNA | BC082098.1 | - | 2, 3 | 856 | 7134452 | |
| FQ229860.1 | - | 2, 3 | 857 | |||