siRNA Details
| Gene Symbol: Fbxo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011998.1 | NP_001011998.1 | 8, 9 | 1101 | ||
| XM_006243419.2 | XP_006243481.1 | 8, 9 | 819 | |||
| GenBank mRNA | BC085831.1 | - | 8, 9 | 1101 | 7134220 | |

Fbxo9
Genomic Map