siRNA Details
| Gene Symbol: Trim43a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142944.1 | NP_001136416.1 | 1 | 20 | ||
| XM_006243512.2 | XP_006243574.1 | 2 | 260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim43a| Gene Symbol: Trim43a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142944.1 | NP_001136416.1 | 1 | 20 | ||
| XM_006243512.2 | XP_006243574.1 | 2 | 260 | |||