siRNA Details
| Gene Symbol: Tmed3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004249.1 | NP_001004249.1 | 3 | 631 | ||
| GenBank mRNA | BC079220.1 | - | 3 | 631 | 7131966 | |
| FQ223087.1 | - | 3 | 569 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmed3| Gene Symbol: Tmed3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004249.1 | NP_001004249.1 | 3 | 631 | ||
| GenBank mRNA | BC079220.1 | - | 3 | 631 | 7131966 | |
| FQ223087.1 | - | 3 | 569 | |||