siRNA Details
| Gene Symbol: Cep63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037772.1 | NP_001032861.2 | 12 | 1523 | ||
| NM_001244805.1 | NP_001231734.1 | 12 | 1692 | |||
| XM_008766490.1 | XP_008764712.1 | 14 | 1814 | |||
| XM_008766491.1 | XP_008764713.1 | 13 | 1538 | |||
| XM_008766492.1 | XP_008764714.1 | 12 | 3113 | |||
| XM_008766493.1 | XP_008764715.1 | 14 | 1689 | |||
| GenBank mRNA | BC083632.1 | - | 15 | 1776 | ||
| BC098948.1 | - | 12 | 1523 | |||
| BC161815.1 | - | 12 | 1692 | 9031816 | ||

Cep63
Genomic Map