siRNA Details
| Gene Symbol: Cep63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037772.1 | NP_001032861.2 | 3 | 216 | ||
| NM_001244805.1 | NP_001231734.1 | 3 | 385 | |||
| XM_008766490.1 | XP_008764712.1 | 5 | 507 | |||
| XM_008766491.1 | XP_008764713.1 | 4 | 231 | |||
| XM_008766492.1 | XP_008764714.1 | 4 | 2040 | |||
| XM_008766493.1 | XP_008764715.1 | 5 | 382 | |||
| GenBank mRNA | BC083632.1 | - | 5 | 375 | ||
| BC098948.1 | - | 3 | 216 | |||
| BC161815.1 | - | 3 | 385 | 9031816 | ||

Cep63
Genomic Map