siRNA Details
| Gene Symbol: Abhd14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009670.1 | NP_001009670.1 | 6 | 1185 | ||
| XM_006243717.2 | XP_006243779.1 | 5 | 1026 | |||
| XM_006243720.2 | XP_006243782.1 | 5 | 1780 | |||
| XM_008766494.1 | XP_008764716.1 | 6 | 1597 | |||
| XM_008766495.1 | XP_008764717.1 | 7 | 1251 | |||
| XM_008766496.1 | XP_008764718.1 | 6 | 1200 | |||
| XM_008766497.1 | XP_008764719.1 | 6 | 1069 | |||
| XM_008766498.1 | XP_008764720.1 | 5 | 1021 | |||
| XM_008766499.1 | XP_008764721.1 | 5 | 894 | |||
| GenBank mRNA | BC088472.1 | - | 6 | 1185 | 7128506 | |

Abhd14a
Genomic Map