siRNA Details
| Gene Symbol: Abhd14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007664.1 | NP_001007665.1 | 4 | 581 | ||
| XM_006243724.1 | XP_006243786.1 | 3 | 522 | |||
| GenBank mRNA | BC076385.1 | - | 4 | 581 | 7108802 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abhd14b| Gene Symbol: Abhd14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007664.1 | NP_001007665.1 | 4 | 581 | ||
| XM_006243724.1 | XP_006243786.1 | 3 | 522 | |||
| GenBank mRNA | BC076385.1 | - | 4 | 581 | 7108802 | |