siRNA Details
| Gene Symbol: Parp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008328.1 | NP_001008329.1 | 2, 3 | 307 | ||
| XM_006243725.2 | XP_006243787.1 | 2, 3 | 628 | |||
| XM_008766500.1 | XP_008764722.1 | 2, 3 | 659 | |||
| GenBank mRNA | BC085863.1 | - | 2, 3 | 307 | 7190424 | |

Parp3
Genomic Map