siRNA Details
| Gene Symbol: Hemk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106853.1 | NP_001100323.1 | 11 | 1194 | ||
| XM_006243728.2 | XP_006243790.1 | 12 | 1484 | |||
| XM_006243729.2 | XP_006243791.1 | 12 | 1804 | |||
| XM_006243730.2 | XP_006243792.1 | 11 | 1559 | |||
| XM_008766502.1 | XP_008764724.1 | 12 | 1419 | |||
| GenBank mRNA | BC161944.1 | - | 11 | 1188 | 9024451 | |

Hemk1
Genomic Map