siRNA Details
| Gene Symbol: Ubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191819.1 | NP_001178748.1 | 12 | 1442 | ||
| XM_006243962.1 | XP_006244024.1 | 11 | 1317 | |||
| XM_006243963.2 | XP_006244025.1 | 12 | 1425 | |||
| XM_008766630.1 | XP_008764852.1 | 13 | 1474 | |||
| XM_008766631.1 | XP_008764853.1 | 11 | 1317 | |||

Ubp1
Genomic Map