siRNA Details
Gene Symbol: Ctdspl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106865.1 | NP_001100335.1 | 7 | 651 | ||
XM_008766675.1 | XP_008764897.1 | 4 | 3517 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ctdspl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106865.1 | NP_001100335.1 | 7 | 651 | ||
XM_008766675.1 | XP_008764897.1 | 4 | 3517 |