siRNA Details
| Gene Symbol: Crb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025661.1 | NP_001020832.1 | 2 | 190 | ||
| XM_006244303.2 | XP_006244365.1 | 2 | 1307 | |||
| GenBank mRNA | BC097382.1 | - | 2 | 190 | 7446098 | |
| FQ223867.1 | - | 423 | ||||

Crb3
Genomic Map