siRNA Details
| Gene Symbol: Crb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025661.1 | NP_001020832.1 | 2, 3 | 231 | ||
| XM_006244303.2 | XP_006244365.1 | 2, 3 | 1348 | |||
| GenBank mRNA | BC097382.1 | - | 2, 3 | 231 | 7446098 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crb3| Gene Symbol: Crb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025661.1 | NP_001020832.1 | 2, 3 | 231 | ||
| XM_006244303.2 | XP_006244365.1 | 2, 3 | 1348 | |||
| GenBank mRNA | BC097382.1 | - | 2, 3 | 231 | 7446098 | |