siRNA Details
| Gene Symbol: Znrf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024878.1 | NP_001020049.1 | 1 | 568 | ||
| GenBank mRNA | BC098025.1 | - | 1 | 568 | 7455918 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znrf4| Gene Symbol: Znrf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024878.1 | NP_001020049.1 | 1 | 568 | ||
| GenBank mRNA | BC098025.1 | - | 1 | 568 | 7455918 | |