siRNA Details
| Gene Symbol: Trem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106884.1 | NP_001100354.1 | 4 | 844 | ||
| XM_006244424.2 | XP_006244486.1 | 4 | 863 | |||
| XM_006244425.2 | XP_006244487.1 | 4 | 860 | |||
| GenBank mRNA | FQ229837.1 | - | 4 | 692 | ||

Trem2
Genomic Map