siRNA Details
| Gene Symbol: Mrpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034136.1 | NP_001029308.1 | 2 | 237 | ||
| GenBank mRNA | BC100081.1 | - | 2 | 237 | 7325700 | |
| FQ214924.1 | - | 2 | 270 | |||
| FQ229593.1 | - | 2 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl2| Gene Symbol: Mrpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034136.1 | NP_001029308.1 | 2 | 237 | ||
| GenBank mRNA | BC100081.1 | - | 2 | 237 | 7325700 | |
| FQ214924.1 | - | 2 | 270 | |||
| FQ229593.1 | - | 2 | 271 | |||