siRNA Details
| Gene Symbol: Mrps18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198756.2 | NP_942051.1 | 3 | 233 | ||
| GenBank mRNA | BC058507.1 | - | 3 | 237 | 6889538 | |
| FQ215521.1 | - | 4 | 412 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps18a| Gene Symbol: Mrps18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198756.2 | NP_942051.1 | 3 | 233 | ||
| GenBank mRNA | BC058507.1 | - | 3 | 237 | 6889538 | |
| FQ215521.1 | - | 4 | 412 | |||