siRNA Details
Gene Symbol: Mrps18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198756.2 | NP_942051.1 | 4 | 361 | ||
GenBank mRNA | BC058507.1 | - | 4 | 365 | 6889538 | |
FQ215521.1 | - | 5 | 540 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mrps18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198756.2 | NP_942051.1 | 4 | 361 | ||
GenBank mRNA | BC058507.1 | - | 4 | 365 | 6889538 | |
FQ215521.1 | - | 5 | 540 |