siRNA Details
| Gene Symbol: Il17f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015011.1 | NP_001015011.1 | 5 | 475 | ||
| GenBank mRNA | BC091568.1 | - | 5 | 475 | 7443410 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il17f| Gene Symbol: Il17f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015011.1 | NP_001015011.1 | 5 | 475 | ||
| GenBank mRNA | BC091568.1 | - | 5 | 475 | 7443410 | |