siRNA Details
| Gene Symbol: Map4k4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106904.1 | NP_001100374.1 | 24 | 2893 | ||
| XM_008767000.1 | XP_008765222.1 | 26 | 3370 | |||
| XM_008767001.1 | XP_008765223.1 | 26 | 3367 | |||
| XM_008767002.1 | XP_008765224.1 | 26 | 3364 | |||
| XM_008767003.1 | XP_008765225.1 | 25 | 3361 | |||
| XM_008767004.1 | XP_008765226.1 | 26 | 3370 | |||
| XM_008767005.1 | XP_008765227.1 | 26 | 3277 | |||
| XM_008767006.1 | XP_008765228.1 | 25 | 3208 | |||
| XM_008767007.1 | XP_008765229.1 | 25 | 3178 | |||
| XM_008767008.1 | XP_008765230.1 | 25 | 3178 | |||
| XM_008767009.1 | XP_008765231.1 | 25 | 3139 | |||
| XM_008767010.1 | XP_008765232.1 | 25 | 3115 | |||
| XM_008767011.1 | XP_008765233.1 | 24 | 3037 | |||
| XM_008767012.1 | XP_008765234.1 | 24 | 2977 | |||
| XM_008767013.1 | XP_008765235.1 | 24 | 2884 | |||
| XM_008767014.1 | XP_008765236.1 | 23 | 2845 | |||
| XM_008767015.1 | XP_008765237.1 | 24 | 2923 | |||
| XM_008767016.1 | XP_008765238.1 | 24 | 2884 | |||
| XM_008767017.1 | XP_008765239.1 | 23 | 2785 | |||

Map4k4
Genomic Map