siRNA Details
| Gene Symbol: Mfsd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106911.1 | NP_001100381.1 | 3 | 1317 | ||
| XM_006244897.2 | XP_006244959.1 | 2 | 2728 | |||
| XM_006244898.2 | XP_006244960.1 | 2 | 2354 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd6| Gene Symbol: Mfsd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106911.1 | NP_001100381.1 | 3 | 1317 | ||
| XM_006244897.2 | XP_006244959.1 | 2 | 2728 | |||
| XM_006244898.2 | XP_006244960.1 | 2 | 2354 | |||