siRNA Details
| Gene Symbol: Fastkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009673.1 | NP_001009673.1 | 3 | 958 | ||
| XM_006245050.1 | XP_006245112.1 | 4 | 1085 | |||
| GenBank mRNA | BC088416.1 | - | 3 | 958 | 7112913 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fastkd2| Gene Symbol: Fastkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009673.1 | NP_001009673.1 | 3 | 958 | ||
| XM_006245050.1 | XP_006245112.1 | 4 | 1085 | |||
| GenBank mRNA | BC088416.1 | - | 3 | 958 | 7112913 | |