siRNA Details
| Gene Symbol: Dnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024879.1 | NP_001020050.1 | 7, 8 | 750 | ||
| XM_006245184.2 | XP_006245246.1 | 7, 8 | 748 | |||
| XM_006245185.2 | XP_006245247.1 | 7, 8 | 849 | |||
| XM_006245186.2 | XP_006245248.1 | 7, 8 | 801 | |||
| GenBank mRNA | BC097388.1 | - | 7, 8 | 750 | 7453069 | |
| FQ229070.1 | - | 4, 5 | 719 | |||
| FQ234491.1 | - | 7, 8 | 657 | |||

Dnpep
Genomic Map