siRNA Details
| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004252.1 | NP_001004252.1 | 15 | 1456 | ||
| XM_006245189.2 | XP_006245251.1 | 15 | 1492 | |||
| GenBank mRNA | BC079364.1 | - | 15 | 1456 | 7109296 | |
| FQ233375.1 | - | 8 | 745 | |||

Farsb
Genomic Map