siRNA Details
| Gene Symbol: Wdfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008331.1 | NP_001008332.1 | 4 | 430 | ||
| XM_006245190.2 | XP_006245252.1 | 5 | 514 | |||
| GenBank mRNA | BC085807.1 | - | 4 | 430 | 7112886 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdfy1| Gene Symbol: Wdfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008331.1 | NP_001008332.1 | 4 | 430 | ||
| XM_006245190.2 | XP_006245252.1 | 5 | 514 | |||
| GenBank mRNA | BC085807.1 | - | 4 | 430 | 7112886 | |