siRNA Details
| Gene Symbol: Itm2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009674.1 | NP_001009674.1 | 2 | 273 | ||
| GenBank mRNA | BC087127.1 | - | 2 | 273 | 7111761 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itm2c| Gene Symbol: Itm2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009674.1 | NP_001009674.1 | 2 | 273 | ||
| GenBank mRNA | BC087127.1 | - | 2 | 273 | 7111761 | |