siRNA Details
| Gene Symbol: Fam174a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012350.1 | NP_001012350.1 | 2 | 610 | ||
| XM_006245579.2 | XP_006245641.1 | 2 | 736 | |||
| GenBank mRNA | BC089835.1 | - | 2 | 610 | 7326083 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam174a| Gene Symbol: Fam174a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012350.1 | NP_001012350.1 | 2 | 610 | ||
| XM_006245579.2 | XP_006245641.1 | 2 | 736 | |||
| GenBank mRNA | BC089835.1 | - | 2 | 610 | 7326083 | |