siRNA Details
| Gene Symbol: Arhgap28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191815.1 | NP_001178744.1 | 12 | 1470 | ||
| XM_006245626.2 | XP_006245688.1 | 12 | 2197 | |||
| XM_006245627.2 | XP_006245689.1 | 12 | 1867 | |||
| XM_006245628.2 | XP_006245690.2 | 12 | 1955 | |||
| XM_008767424.1 | XP_008765646.1 | 12 | 1377 | |||
| XM_008767425.1 | XP_008765647.1 | 12 | 1712 | |||

Arhgap28
Genomic Map